Anti-PRRT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17261-1-AP-150UL
10092-852EA 402 USD
10092-852
Anti-PRRT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF:transfected Cells, N/A

Type: Primary
Antigen: PRRT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR