Anti-MRAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17259-1-AP-150UL
10090-406EA 402 USD
10090-406
Anti-MRAP2 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: MRAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR