Anti-ATPB Rabbit Polyclonal Antibody

Supplier: Proteintech
17247-1-AP-150UL
10083-390EA 402 USD
10083-390
Anti-ATPB Rabbit Polyclonal Antibody
Antibodies
ATP5B, also named as ATPMB and ATPSB, belongs to the ATPase alpha/beta chains family. It is a high affinity HDL receptor for apolipoprotein A1. ATP5B is a subunit of mitochondrial ATP synthase (F1F0 ATP synthase or Complex V ).

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200; IP: Mouse Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: ATPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR