Anti-HLA-DRA Rabbit Polyclonal Antibody

Supplier: Proteintech
17221-1-AP-150UL
10088-302EA 402 USD
10088-302
Anti-HLA-DRA Rabbit Polyclonal Antibody
Antibodies
IHC: Human tonsil, 1:20-1:200

Type: Primary
Antigen: HLA-DRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR