Anti-LYZL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17207-1-AP-150UL
10089-844EA 402 USD
10089-844
Anti-LYZL1 Rabbit Polyclonal Antibody
Antibodies
LYZL1(Lysozyme-like 1) is also named as LYC2 and belongs to the glycosyl hydrolase 22 family. It is involved in the hydrolysis of 1,4-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in a peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrins. LYZL1 has two isoforms with the molecular weight of 17 kDa and 22 kDa. The full length protein has a signal peptide of 19 amino acids and a glycosylation site. This antibody detects the N-terminal of LYZL1.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: LYZL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR