Anti-RNASE11 Rabbit Polyclonal Antibody

Supplier: Proteintech
17203-1-AP-150UL
10093-764EA 402 USD
10093-764
Anti-RNASE11 Rabbit Polyclonal Antibody
Antibodies
RNASE11, also named as C14ORF6, is ribonuclease 11. This antibody detected 30 kDa band with Testis and L02 cell lysate.

Western Blot: Human Testis Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: RNASE11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR