Anti-KCNN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
17188-1-AP-150UL
10088-196EA 402 USD
10088-196
Anti-KCNN3 Rabbit Polyclonal Antibody
Antibodies
IF:SH-SY5Y Cells, 1:10-1:100; Western Blot:Transfected HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: KCNN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR