Anti-Transition Protein 1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17178-1-AP-150UL
10096-162EA 402 USD
10096-162
Anti-Transition Protein 1 Rabbit Polyclonal Antibody
Antibodies
Transition nuclear proteins (TNP1 and TNP2) are the major nuclear proteins that replace somatic histones during spermatogenesis. TNPs are required for normal chromatin condensation and functional sperm development, spermatogenesis was found to be compromised in both Tnp1 and Tnp2 mice. TNP1, or TP1, localized in nucleus, is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Recently, TNP-1 was used as a germ cell marker in condensing spermatids.

Western Blot: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: Transition Protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR