Anti-IARS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17170-1-AP-150UL
10088-554EA 537.7 USD
10088-554
Anti-IARS2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Brain, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: IARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR