Anti-Histone-H3 Rabbit Polyclonal Antibody

Supplier: Proteintech
17168-1-AP-150UL
10088-282EA 509.69 USD
10088-282
Anti-Histone-H3 Rabbit Polyclonal Antibody
Antibodies
HIST2H3A,histone cluster 2, H3a.It is the core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. HIST2H3A is Expressed during S phase, then expression strongly decreases as cell division slows down during the process of differentiation.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human skin cancer Tissue, 1:20-1:200; IF: HEK-293 Cells, 1:20-1:200

Type: Primary
Antigen: Histone-H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR