Anti-ACAD10 Rabbit Polyclonal Antibody

Supplier: Proteintech
17161-1-AP-150UL
10082-384EA 402 USD
10082-384
Anti-ACAD10 Rabbit Polyclonal Antibody
Antibodies
ACAD10(Acyl-CoA dehydrogenase family member 10) belongs to the acyl-CoA dehydrogenase family. ACAD10 has significant activity towards the branched-chain substrates R and S, 2 methyl-C15-CoA and is highly expressed in fetal but not adult brain. This pattern of expression is similar to that of LCAD, another ACAD previously shown to be involved in long branched chain fatty acid metabolism. It has 4 isoforms produced by alternative splicing.

Western Blot: Mouse spleen Tissue, 1:200-1:2000; IHC: Human pancreas Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: ACAD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR