Anti-FCRLB Rabbit Polyclonal Antibody

Supplier: Proteintech
17157-1-AP-150UL
10086-924EA 402 USD
10086-924
Anti-FCRLB Rabbit Polyclonal Antibody
Antibodies
IHC: Human lung cancer Tissue, 1:20-1:200; Western Blot:Transfected HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: FCRLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR