Anti-FAM160B2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17147-1-AP-150UL
10086-800EA 402 USD
10086-800
Anti-FAM160B2 Rabbit Polyclonal Antibody
Antibodies
FAM160B2, also named as RAI16, may play a role in early lens development in mammals and in upregulation of genes in CD34+ cells in coronary artery disease patients.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: FAM160B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR