Anti-RBM12B Rabbit Polyclonal Antibody

Supplier: Proteintech
17137-1-AP-150UL
10093-390EA 402 USD
10093-390
Anti-RBM12B Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: RBM12B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR