Anti-OMA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17116-1-AP-150UL
10091-582EA 524.48 USD
10091-582
Anti-OMA1 Rabbit Polyclonal Antibody
Antibodies
OMA1(Overlapping with the m-AAA protease 1 homolog) is also named as MPRP1(Metalloprotease-related protein 1) and belongs to the peptidase M48 family.It is a zinc metalloprotease that spans the inner mitochondrial membrane with a number of predicted membrane spanning domains and the 60 kDa form of OMA1 rapidly accumulated concomitantly with the cleavage of all OPA1 variants,and the 40 kDa form of OMA1 may be the inactive form, with the 60 kDa form being the active enzyme.This antibody is speicific to OMA1.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: OMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR