Anti-ATP6V1A Rabbit Polyclonal Antibody

Supplier: Proteintech
17115-1-AP-150UL
10081-954EA 402 USD
10081-954
Anti-ATP6V1A Rabbit Polyclonal Antibody
Antibodies
The vacuolar-type H(+)-ATPase (V-ATPase) is responsible for the acidification of endosomes, lysosomes, and other intracellular organelles. It is also involved in hydrogen ion transport across the plasma membrane into the extracellular space. The V-ATPase is a multisubunit complex with cytosolic and transmembrane domains. The cytosolic catalytic domain consists of 3 A subunits and 3 B subunits, which bind and hydrolyze ATP, as well as regulatory accessory subunits.ATP6V1A is V-type proton ATPase catalytic subunit A.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human pancreas, 1:20-1:200

Type: Primary
Antigen: ATP6V1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR