Anti-GNPDA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17105-1-AP-150UL
10087-718EA 402 USD
10087-718
Anti-GNPDA2 Rabbit Polyclonal Antibody
Antibodies
GNPDA2(Glucosamine-6-phosphate deaminase 2) is also named as GNP2 and belongs to the glucosamine/galactosamine-6-phosphate isomerase family. It catalyzes the reversible conversion of D-glucosamine-6-phosphate into D-fructose-6-phosphate and ammonium. Human GNPDA1 and GNPDA2 shared the same fold and were predicted to form hexameric particles. This protein has 5 isoforms produced by alternative splicing with the molecular weight of 31 kDa, 29 kDa, 23 kDa and 27 kDa.

Western Blot:rat Kidney Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GNPDA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR