Anti-LARS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17097-1-AP-150UL
10089-524EA 524.48 USD
10089-524
Anti-LARS2 Rabbit Polyclonal Antibody
Antibodies
LARS2(Probable leucine--tRNA ligase, mitochondrial) is also named as mitochondrial LEURS. The deduced 903-amino acid protein has an N-terminal mitochondrial targeting signal that is cleaved after residue 39. LARS2 has characteristics of a class I aminoacyl-tRNA synthetase, including a classical Rossmann fold. It belongs to the class-I aminoacyl-tRNA synthetase family.

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: LARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR