Anti-MFF Rabbit Polyclonal Antibody

Supplier: Proteintech
17090-1-AP-150UL
10090-176EA 537.7 USD
10090-176
Anti-MFF Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: MFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR