Anti-RPL24 Rabbit Polyclonal Antibody

Supplier: Proteintech
17082-1-AP-150UL
10093-538EA 524.48 USD
10093-538
Anti-RPL24 Rabbit Polyclonal Antibody
Antibodies
The mammalian ribosome comprises 79 ribosomal proteins and four rRNAs, which combine in equimolar ratios to form the small (40S) and large (60S) subunits. Ribosome proteins are a direct and critical target of the PI3K pathway in promoting growth.. RPL24 is one component of the large (60S) subunits that promote the translation of uORF-containing mRNAsgene The mutation in Rpl24 result in impairment of mRNA splicing and L24 production, which in turn affects ribosome biogenesis, protein synthesis and the cell cycle. PMID:20799971]. Also RPL24 (ribosomal protein L24) is a key factor for translation reinitiation of downstream ORFs on the polycistronic cauliflower mosaic virus 35S RNA transcription unit, and may have a role in gynoecuim development.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Placenta, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: RPL24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR