Anti-CST6 Rabbit Polyclonal Antibody

Supplier: Proteintech
17076-1-AP-150UL
10085-272EA 402 USD
10085-272
Anti-CST6 Rabbit Polyclonal Antibody
Antibodies
IHC: Mouse Liver, 1:20-1:200

Type: Primary
Antigen: CST6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR