Anti-ZNF18 Rabbit Polyclonal Antibody

Supplier: Proteintech
17070-1-AP-150UL
10097-310EA 402 USD
10097-310
Anti-ZNF18 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: ZNF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR