Anti-EIF5A2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17069-1-AP-150UL
10086-454EA 402 USD
10086-454
Anti-EIF5A2 Rabbit Polyclonal Antibody
Antibodies
Eukaryotic initiation factor 5A (EIF5A) plays an essential role in the viability of eukaryotic cells. EIF5A is known to act as a translation initiation factor specific for a small number of mRNAs, a cellular target of HIV-1 REV protein, and an exportin-4-dependent nuclear export cargo. It is also involved in mRNA turnover and the establishment of actin polarity. . EIF5A2, one isoform of EIF5A, has a key at the level of mRNA turnoverby acting downstream of decapping. It also involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity .

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: EIF5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR