Anti-ARL8A Rabbit Polyclonal Antibody

Supplier: Proteintech
17060-1-AP-150UL
10082-672EA 537.7 USD
10082-672
Anti-ARL8A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: ARL8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR