Anti-STARD10 Rabbit Polyclonal Antibody

Supplier: Proteintech
17048-1-AP-150UL
10095-246EA 402 USD
10095-246
Anti-STARD10 Rabbit Polyclonal Antibody
Antibodies
STARD10 (START domain-containing protein 10; also known as PCTP-like protein) was firstly identified as a 40 kDa protein associated with the maturation of sperm (10819773). It is highly expressed in liver and responsible to transfer phosphatidylcholine. STARD10 has also been reported to be overexpressed in breast tumors, suggesting its possible role in tumorigenesis. This antibody detected duplicate bands between 35-40 kDa in MCF7 and HepG2 cells. The similar expression pattern of STARD10 was also observed by other scientists, while the exact reason is not clear (15150109).

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: STARD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR