Anti-GALT Rabbit Polyclonal Antibody

Supplier: Proteintech
17035-1-AP-150UL
10087-366EA 402 USD
10087-366
Anti-GALT Rabbit Polyclonal Antibody
Antibodies
GALT (galactose-1-phosphate uridylyltransferase) catalyses the second step in the Leloir pathway for the conversion of galactose-l-phosphate (Gal-I-P) into uridine diphosphogalactose (UDP-gal). Defects in GALT cause galactosemia.

Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: GALT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR