Anti-GRK5 Rabbit Polyclonal Antibody

Supplier: Proteintech
17032-1-AP-150UL
10087-876EA 402 USD
10087-876
Anti-GRK5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: GRK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR