Anti-GEFT Rabbit Polyclonal Antibody

Supplier: Proteintech
17028-1-AP-150UL
10087-480EA 402 USD
10087-480
Anti-GEFT Rabbit Polyclonal Antibody
Antibodies
GEFT, also named as p63RhoGEF, may play a role in actin cytoskeleton reorganization in different tissues since its activation induces formation of actin stress fibers. It works as a guanine nucleotide exchange factor for Rho family of small GTPases. GETF links specifically G alpha q/11-coupled receptors to RHOA activation. GEFT may be an important regulator of processes involved in axon and dendrite formation. In neurons seems to be an exchange factor primarily for RAC1. It is involved in skeletal myogenesis.

Western Blot: Human Brain Tissue, 1:200-1:1000; IP: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: GEFT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR