Anti-Collagen Type VI Rabbit Polyclonal Antibody

Supplier: Proteintech
17023-1-AP-150UL
10084-996EA 402 USD
10084-996
Anti-Collagen Type VI Rabbit Polyclonal Antibody
Antibodies
Type VI Collagen is a major structural component of microfibrils. Type VI Collagen was found to be present throughout the connective tissue and in the extracellular matrix of cultured fibroblasts . It is a heterotrimer composed of three different chains: alpha-1(VI), alpha-2(VI), and alpha-3(VI) or alpha-5(VI) or alpha-6(VI). The alpha-1(VI) chain is encoded by COL6A1 gene. Mutations in this gene result in Bethlem myopathy and Ullrich congenital muscular dystrophy.

Western Blot: Human Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: Mouse Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: Collagen Type VI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR