Anti-FAM65B Rabbit Polyclonal Antibody

Supplier: Proteintech
17015-1-AP-150UL
10086-830EA 402 USD
10086-830
Anti-FAM65B Rabbit Polyclonal Antibody
Antibodies
FAM65B is also named as C6orf32, DIFF48, KIAA0386, PL48 and belongs to the FAM65 family.It has 2 isoforms produced by alternative splicing.Isoform 2 play a role in promoting myogenic cell differentiation, cytoskeletal rearrangement and filopodia formation. FAM65B is expressed in mononuclear cells, not in myotubes, upon induction of differentiation and cellular fusion, and expression is upregulated during human myoblast differentiation at both the mRNA and protein levels.This antibody is specific to FAM65B.

Western Blot: Human Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: FAM65B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR