Anti-HPD Rabbit Polyclonal Antibody

Supplier: Proteintech
17004-1-AP-150UL
10088-424EA 402 USD
10088-424
Anti-HPD Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: HPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR