Anti-METTL7B Rabbit Polyclonal Antibody

Supplier: Proteintech
17001-1-AP-150UL
10090-168EA 537.7 USD
10090-168
Anti-METTL7B Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: METTL7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR