Anti-SPATA22 Rabbit Polyclonal Antibody

Supplier: Proteintech
16989-1-AP-150UL
10095-076EA 402 USD
10095-076
Anti-SPATA22 Rabbit Polyclonal Antibody
Antibodies
Spata22 (NYD-SP20), a novel vertebrate-specific gene, is required for meiotic progress in mouse germ cells. It is essential for one or more key events of early meiotic prophase, as homologous chromosomes of mutant germ cells do not achieve normal synapsis or repair meiotic DNA double-strand breaks.

Western Blot: Human Testis Tissue, 1:200-1:2000; IP: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: SPATA22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR