Anti-PHYHIP Rabbit Polyclonal Antibody

Supplier: Proteintech
16984-1-AP-150UL
10092-218EA 402 USD
10092-218
Anti-PHYHIP Rabbit Polyclonal Antibody
Antibodies
PHYHIP, also named as DYRK1AP3 and KIAA0273, belongs to the PHYHIP family. Its interaction with PHYH suggests a role in the development of the central system. This antibody recognizes both PHYHIP (35-38 kDa) and PHYHIPL (43-45 kDa).

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: PHYHIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR