Anti-NVL Rabbit Polyclonal Antibody

Supplier: Proteintech
16970-1-AP-150UL
10091-496EA 402 USD
10091-496
Anti-NVL Rabbit Polyclonal Antibody
Antibodies
NVL(nuclear VCP-like) belongs to the AAA ATPase family and may facilitate the dissociation and recycling of nucleolin, thereby promoting efficient ribosome biogenesis. It acts as an ATP-dependent zinc metallopeptidase. This protein has 5 isoforms produced by alternative splicing.

Western Blot: Mouse lung Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: NVL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR