Anti-CA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16961-1-AP-150UL
10083-916EA 537.7 USD
10083-916
Anti-CA2 Rabbit Polyclonal Antibody
Antibodies
CA2(carbonic anhydrase II ) is also named as Car2, CA-II, CAII and belongs to the alpha-carbonic anhydrase family. It reversibly hydrates CO2 in cellular ion transport and homeostasis. CA2 is essential for bone resorption and osteoclast differentiation. It regulates SLC9A1 activity via a phosphorylation-regulated CAII binding site in SLC9A1 tail. It also may be involved in the development of brain. Defects in CA2 are the cause of osteopetrosis autosomal recessive type 3 (OPTB3).

Western Blot: Mouse ovary Tissue, 1:500-1:5000

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR