Anti-CPLX3 Rabbit Polyclonal Antibody

Supplier: Proteintech
16949-1-AP-150UL
10085-114EA 402 USD
10085-114
Anti-CPLX3 Rabbit Polyclonal Antibody
Antibodies
CPLX3, also named as Complexin-3, is 158 amino acid protein, which belongs to the complexin/synaphin family. CPLX3 localizes in the membrane and binds to the SNARE core complex containing SNAP25, VAMP2 and STX1A. CPLX3 positively regulates a late step in synaptic vesicle exocytosis.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: CPLX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR