Anti-RPS21 Rabbit Polyclonal Antibody

Supplier: Proteintech
16946-1-AP-150UL
10093-952EA 402 USD
10093-952
Anti-RPS21 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: RPS21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR