Anti-RNF10 Rabbit Polyclonal Antibody

Supplier: Proteintech
16936-1-AP-150UL
10093-782EA 402 USD
10093-782
Anti-RNF10 Rabbit Polyclonal Antibody
Antibodies
RING finger (C3HC4-type zinc finger) is a variant zinc finger motif present in a new family of proteins including transcription regulators. RNF10 (RING finger protein 10), a 90-kDa protein, contains a ring finger motif which is known to be involved in protein-protein interactions. RNF10 regulates myelin-associated glycoprotein (MAG) expression and is required for myelin formation, therefore, participates in the peripheral nerve development and Schawnn cell differentiation. RNF10 is also involved in activation of p21WAF1 promoter via its activity as transcriptional factor.

Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: RNF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR