Anti-AP1B1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16932-1-AP-150UL
10082-958EA 402 USD
10082-958
Anti-AP1B1 Rabbit Polyclonal Antibody
Antibodies
AP1B1 is a subunit of the adaptor complex AP-1 and is a member of the adaptin protein family. Adaptor protein (AP) complexes are cytosolic heterotetramers that mediate the sorting of membrane proteins in the secretory and endocytic pathways. AP-1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors.

Western Blot: HEK-293 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: AP1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR