Anti-PRPF40B Rabbit Polyclonal Antibody

Supplier: Proteintech
16929-1-AP-150UL
10092-834EA 402 USD
10092-834
Anti-PRPF40B Rabbit Polyclonal Antibody
Antibodies
Western Blot:Y79 Cells, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: PRPF40B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR