Anti-CAPS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16926-1-AP-150UL
10083-970EA 402 USD
10083-970
Anti-CAPS1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Colon Tissue, 1:200-1:2000

Type: Primary
Antigen: CAPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR