Anti-HSD17B7 Rabbit Polyclonal Antibody

Supplier: Proteintech
16925-1-AP-150UL
10081-860EA 402 USD
10081-860
Anti-HSD17B7 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200; IP:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: HSD17B7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR