Anti-ATL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
16921-1-AP-150UL
10083-264EA 402 USD
10083-264
Anti-ATL3 Rabbit Polyclonal Antibody
Antibodies
IF: HepG2 Cells, 1:10-1:100; Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: ATL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR