Anti-FAM156A Rabbit Polyclonal Antibody

Supplier: Proteintech
16914-1-AP-150UL
10086-798EA 402 USD
10086-798
Anti-FAM156A Rabbit Polyclonal Antibody
Antibodies
IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: FAM156A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR