Anti-PROS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16910-1-AP-150UL
10092-814EA 402 USD
10092-814
Anti-PROS1 Rabbit Polyclonal Antibody
Antibodies
PROS1, also named as Vitamin K-dependent protein S, is a 676 amino acid protein, which contains 4 EGF-like domains, contains one Gla (gamma-carboxy-glutamate) domain and contains 2 laminin G-like domains. PROS1 as a secreted protein platelets alpha granules is expressed in plasma. PROS1 is a cofactor to activated protein C in the degradation of coagulation factors Va and VIIIa and helps to prevent coagulation and stimulating fibrinolysis.

Western Blot: Human Placenta Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: PROS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR