Anti-GPT Rabbit Polyclonal Antibody

Supplier: Proteintech
16897-1-AP-150UL
10087-816EA 402 USD
10087-816
Anti-GPT Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: GPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR