Anti-RPGR Rabbit Polyclonal Antibody

Supplier: Proteintech
16891-1-AP-150UL
10093-890EA 537.7 USD
10093-890
Anti-RPGR Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Eye Tissue, 1:200-1:2000; IF: HTERT-RPE1 Cells, 1:10-1:100; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: RPGR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR