Anti-Pepsinogen II Rabbit Polyclonal Antibody

Supplier: Proteintech
16881-1-AP-150UL
10092-142EA 402 USD
10092-142
Anti-Pepsinogen II Rabbit Polyclonal Antibody
Antibodies
Pepsinogen Iialso known as progastricsin or PGC(pepsinogen C), is an aspartic protease expressed primarily in gastric chief cells and is a novel marker of type 2 cells with advantages over many of the current markers used to identify type 2 cells in the developing lung. It is involved in proteolysis and peptidolysis. This protein has 2 isoforms produced by alternative splicing.

Western Blot: Human Stomach Tissue, 1:200-1:2000

Type: Primary
Antigen: Pepsinogen II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR