Anti-FGF16 Rabbit Polyclonal Antibody

Supplier: Proteintech
16876-1-AP-150UL
10086-972EA 402 USD
10086-972
Anti-FGF16 Rabbit Polyclonal Antibody
Antibodies
FGF16 is a member of the fibroblast growth factor (FGF) family. Fibroblast growth factor (Fgf) signaling plays important roles in development and metabolism. FFGF16 is predominantly expressed in the heart. It plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation, and is required for normal cardiomyocyte proliferation and heart development. This antibody is specific to FGF16.

IHC: Human Heart Tissue, 1:20-1:200; Western Blot: Human Heart Tissue, 1:200-1:1000

Type: Primary
Antigen: FGF16-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR